2,2,2-Trifluoroethanol-[D₃] (99% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
42363-03
42363-46
CAAA42363-03EA
244.9
CAD
CAAA42363-03
CAAA42363-46
2,2,2-Trifluoroethanol-[D₃] (99% D) (Isotopic) for NMR spectroscopy
2,2,2-Trifluoroethanol-[D3]
NMR Solvent
Purity: 99%
CAS No.: 77253-67-9
Molecular Formula: CF3CD2OD
Formula Weight: 103.06
Physical Form: Liquid in prescored ampoules
Density: 1.45
Melting Point: -44°
Boiling Point: 77-80°
UN No.: 1992
MDL No.: MFCD00037682
NMR Solvent
Formula:
F₃CCD₂OD Boiling Pt: 77…80 °C Melting Pt: –44 °C Density: 1.45 Storage Temperature: Ambient |
MDL Number:
MFCD00037682 CAS Number: 77253-67-9 EINECS: 278-649-6 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...