Molybdenum ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
42361-09
42361-18
CAAA42361-09EA
79.32
CAD
CAAA42361-09
CAAA42361-18
Molybdenum ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Molybdenum
MDL: MFCD00003465
Purity: 99.95%
CAS No.: 7439-98-7
Physical Form: ≈0.26g/slug
MDL No.: MFCD00003465
Formula:
Mo Boiling Pt: 3617 °C (1013 hPa) Melting Pt: 2622 °C Density: 10.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 Merck Index: 12,06317 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...