Trifluoroacetic acid-[D₁] (99.5% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trifluoroacetic [2H]acid, TFA-D, Trifluoroacetic acid-[²H₁]
42357-09
42357-14
CAAA42357-09EA
98.59
CAD
CAAA42357-09
CAAA42357-14
Trifluoroacetic acid-[D₁] (99.5% D) (Isotopic) for NMR spectroscopy
Trifluoroacetic acid-[D1]
MDL: MFCD00044563
Purity: 99.5%
CAS No.: 599-00-8
Molecular Formula: CF3CO2D
Formula Weight: 115.03
Physical Form: Liquid in prescored ampoules
Density: 1.50
Melting Point: -15°
Boiling Point: 72°
UN No.: 2699
MDL No.: MFCD00044563
Formula:
F₃CCOOD Boiling Pt: 72 °C Melting Pt: –15 °C Density: 1.50 Storage Temperature: Ambient |
MDL Number:
MFCD00044563 CAS Number: 599-00-8 EINECS: 209-961-2 UN: 2699 ADR: 8,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...