Cobalt ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42355-06
42355-14
42355-22
CAAA42355-06EA
188.02
CAD
CAAA42355-06
CAAA42355-14
CAAA42355-22
Cobalt ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Cobalt
MDL: MFCD00010935
Purity: 99.95%
CAS No.: 7440-48-4
Physical Form: ≈0.22g/slug
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...