1,1,2,2-Tetrachloroethane-[D₂] (99.6% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,2,2-Tetrachloroethane-[²H₂]

42347-04 42347-09 42347-18
CAAA42347-04EA 65.73 CAD
CAAA42347-04 CAAA42347-09 CAAA42347-18
1,1,2,2-Tetrachloroethane-[D₂] (99.6% D) (Isotopic) for NMR spectroscopy
1,1,2,2-Tetrachloroethane-[D2]

MDL: MFCD00037672


Purity: 99.6% CAS No.: 33685-54-0 Molecular Formula: C2D2Cl4 Formula Weight: 169.86 Physical Form: Liquid Density: 1.620 Refractive Index: 1.4930 Appearance: Colorless Melting Point: -44° Boiling Point: 147° UN No.: 1702 MDL No.: MFCD00037672

Formula: Cl₂CDCDCl₂
Boiling Pt: 147 °C
Melting Pt: –44 °C
Density: 1.620
Storage Temperature: Ambient
MDL Number: MFCD00037672
CAS Number: 33685-54-0
EINECS: 251-634-1
UN: 1702
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR