n-Octane-[D₁₈] (99% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
42339-03
42339-06
CAAA42339-03EA
319.16
CAD
CAAA42339-03
CAAA42339-06
n-Octane-[D₁₈] (99% D) (Isotopic) for NMR spectroscopy
n-Octane-[D18]
MDL: MFCD00037626
Purity: 99%
CAS No.: 17252-77-6
Molecular Formula: CD3(CD2)6CD3
Formula Weight: 132.34
Flash Point: 15.6°(60°F)
Physical Form: Liquid
Density: 0.815
Melting Point: -57°
Boiling Point: 126°
UN No.: 1262
MDL No.: MFCD00037626
Formula:
D₃C(CD₂)₆CD₃ Boiling Pt: 126 °C Melting Pt: –57 °C Density: 0.815 Flash Pt: 15.6 °C (60 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00037626 CAS Number: 17252-77-6 EINECS: 241-285-3 UN: 1262 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...