Dichloromethane-[D₂] (99.9% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
42335-KV
42335-SA
CAAA42335-KVEA
377.62
CAD
CAAA42335-KV
CAAA42335-SA
Dichloromethane-[D₂] (99.9% D) (Isotopic) for NMR spectroscopy
Dichloromethane-[D2]
MDL: MFCD00000882
Purity: 99.9%
CAS No.: 1665-00-5
Molecular Formula: CD2Cl2
Formula Weight: 86.95
Physical Form: Liquid, in prescored ampoules, 0.75ml/ampoule
Density: 1.35
UN No.: 1593
MDL No.: MFCD00000882
Formula:
CD₂Cl₂ Boiling Pt: 40 °C (1013 hPa) Melting Pt: –97 °C Density: 1.35 Storage Temperature: Ambient |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...