Aluminium ≥99.99% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
42328-09
42328-18
42328-30
CAAA42328-09EA
51.82
CAD
CAAA42328-09
CAAA42328-18
CAAA42328-30
Aluminium ≥99.99% (metals basis), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Aluminium
MDL: MFCD00134029
Purity: 99.99%
CAS No.: 7429-90-5
Physical Form: ≈1.1g/slug
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...