Aluminium ≥99.999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
42324-09
42324-18
42324-30
CAAA42324-09EA
61.62
CAD
CAAA42324-09
CAAA42324-18
CAAA42324-30
Aluminium ≥99.999% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Aluminium
MDL: MFCD00134029
Purity: 99.999%
CAS No.: 7429-90-5
Physical Form: ≈1.1g/slug
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...