Methanol-[D₄] (100% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

42320-KV 42320-SA
CAAA42320-KVEA 535.62 CAD
CAAA42320-KV CAAA42320-SA
Methanol-[D₄] (100% D) (Isotopic) for NMR spectroscopy
Methanol-[D4]

MDL: MFCD00044637 Notes: Minimum isotopic purity 99.96%


Purity: 100% CAS No.: 811-98-3 Molecular Formula: CD3OD Formula Weight: 36.07 Flash Point: 11°(52°F) Physical Form: Liquid, in prescored ampoules, 0.75ml/ampoule Density: 0.89 Appearance: Colorless Odor: Mild Melting Point: -98° Boiling Point: 64.7° UN No.: 1230 MDL No.: MFCD00044637

Formula: D₃COD
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.89
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR