Methanol-[D₄] (99.8% D) (Isotopic) + 0.05% v/v TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
42319-KV
42319-SA
CAAA42319-KVEA
273.34
CAD
CAAA42319-KV
CAAA42319-SA
Methanol-[D₄] (99.8% D) (Isotopic) + 0.05% v/v TMS for NMR spectroscopy
Methanol-[D4]
MDL: MFCD00044637
Purity: 99.8%
CAS No.: 811-98-3
Molecular Formula: CD4O
Formula Weight: 36.07
Physical Form: Liquid, in prescored ampoules, 1g/ampoule
Density: 0.89
Appearance: Colorless
Odor: Mild
Melting Point: -98°
Boiling Point: 64.7°
UN No.: 1230
MDL No.: MFCD00044637
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 Melting Pt: -99 Density: 0.89 Flash Pt: 11 Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...