Silver ≥99.998% (metals basis), foil, Premion®, annealed, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
42317-FI
42317-GH
42317-HP
CAAA42317-FIEA
98.28
CAD
CAAA42317-FI
CAAA42317-GH
CAAA42317-HP
Silver ≥99.998% (metals basis), foil, Premion®, annealed, Thickness: 0.1 mm (0.004 in)
Silver
MDL: MFCD00003397
Purity: 99.998%
CAS No.: 7440-22-4
Physical Form: ≈2.6g/50x50mm, Temper: soft
Odor: Characteristic
MDL No.: MFCD00003397
Formula:
Ag Boiling Pt: 2212 °C (1013 hPa) Melting Pt: 962 °C Density: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...