2-Propanol-[D₈] (99% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
IPA-D8, 2-Propanol-[²H₈], 2-Hydroxy propane-D8, Isopropyl alcohol-D8, Isopropanol-D8, Dimethyl carbinol-D8
42314-06
42314-14
CAAA42314-06EA
271.76
CAD
CAAA42314-06
CAAA42314-14
2-Propanol-[D₈] (99% D) (Isotopic) for NMR spectroscopy
2-Propanol-[D8]
MDL: MFCD00044341
Purity: 99%
CAS No.: 22739-76-0
Molecular Formula: (CD3)2CDOD
Formula Weight: 68.15
Flash Point: -11.7°(53°F)
Physical Form: Liquid
Density: 0.90
Odor: Alcohol-like
Melting Point: -89.5°
Boiling Point: 82.4°
UN No.: 1219
MDL No.: MFCD00044341
Formula:
D₃CCD(OD)CD₃ Boiling Pt: 82.4 °C Melting Pt: –89.5 °C Density: 0.90 Flash Pt: –11.7 °C (11 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00044341 CAS Number: 22739-76-0 EINECS: 245-189-2 UN: 1219 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...