1,1,1,3,3,3-Hexafluoro-2-propanol-[D₂] (98% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
HFIP-D2, 1,1,1,3,3,3-Hexafluoroisopropanol-d2, 1,1,1,3,3,3-Hexafluoro-2-propanol-[²H₂]
42291-03
42291-46
CAAA42291-03EA
260.7
CAD
CAAA42291-03
CAAA42291-46
1,1,1,3,3,3-Hexafluoro-2-propanol-[D₂] (98% D) (Isotopic) for NMR spectroscopy
1,1,1,3,3,3-Hexafluoro-2-propanol-[D2]
Purity: 98%
CAS No.: 38701-74-5
Molecular Formula: CF3CD(OD)CF3
Formula Weight: 170.05
Physical Form: Liquid, packaged in prescored ampoules
Density: 1.60
Melting Point: -40°
Boiling Point: 59°
UN No.: 1760
MDL No.: MFCD00037559
Formula:
F₃CCD(OD)CF₃ MW: 170.02 g/mol Boiling Pt: 59 °C Melting Pt: –4 °C Density: 1.60 Storage Temperature: Ambient |
MDL Number:
MFCD00037559 CAS Number: 38701-74-5 EINECS: 254-091-9 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...