Dimethyl sulfoxide-[D₆] (100% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Warning
|
42285-KV
42285-SA
CAAA42285-KVEA
346.02
CAD
CAAA42285-KV
CAAA42285-SA
Dimethyl sulfoxide-[D₆] (100% D) (Isotopic) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
MDL: MFCD00002090
Notes: Minimum isotopic purity 99.96%
Purity: 100%
CAS No.: 2206-27-1
Molecular Formula: (CD3)2SO
Formula Weight: 84.17
Flash Point: 88°(190°F)
Physical Form: Liquid, in prescored ampoules, 0.75ml/ampoule
Density: 1.18
Refractive Index: 1.4760
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Characteristic
Melting Point: 20°
Boiling Point: 190°
MDL No.: MFCD00002090
Formula:
D₃CS(O)CD₃ Boiling Pt: 190 °C Melting Pt: 20 °C Density: 1.18 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002090 CAS Number: 2206-27-1 EINECS: 218-617-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...