N,N-Dimethylformamide-[D₇] (99.5% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
42284-03
42284-46
042284-06
042284-09
CAAA42284-03EA
124.66
CAD
CAAA42284-03
CAAA42284-46
AA42284-06
AA42284-09
N,N-Dimethylformamide-[D₇] (99.5% D) (Isotopic) for NMR spectroscopy
N,N-Dimethylformamide-[D7]
MDL: MFCD00003286
Purity: 99.5%
CAS No.: 4472-41-7
Molecular Formula: C3D7NO
Formula Weight: 80.14
Flash Point: 58°(136°F)
Physical Form: Liquid in prescored ampoules
Density: 1.04
Appearance: Colorless
Odor: Amine-like
Boiling Point: 153°
UN No.: 2265
MDL No.: MFCD00003286
Formula:
DCOON(CD₃)₂ Boiling Pt: 153 °C Melting Pt: –60 °C Density: 1.04 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003286 CAS Number: 4472-41-7 EINECS: 224-745-8 UN: 2265 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...