Cyclohexane-[D₁₂] (99.5% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
42271-KV
42271-SA
CAAA42271-KVEA
622.52
CAD
CAAA42271-KV
CAAA42271-SA
Cyclohexane-[D₁₂] (99.5% D) (Isotopic) for NMR spectroscopy
Cyclohexane-[D12]
MDL: MFCD00044212
Beilstein Registry No.: 1908842
Purity: 99.5%
CAS No.: 1735-17-7
Molecular Formula: C6D12
Formula Weight: 96.24
Flash Point: -18°(-1°F)
Physical Form: Liquid in prescored ampoules, 1g/ampoule
Density: 0.89
Refractive Index: 1.4210
Melting Point: 6.5°
Boiling Point: 81°
Beilstein Reference: 1908842
UN No.: 1145
MDL No.: MFCD00044212
Formula:
C₆D₁₂ |
MDL Number:
MFCD00044212 CAS Number: 1735-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...