Bromobenzene-[D₅] (99% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

42269-04 42269-09
CAAA42269-04EA 103.17 CAD
CAAA42269-04 CAAA42269-09
Bromobenzene-[D₅] (99% D) (Isotopic) for NMR spectroscopy
Bromobenzene-[D5]

MDL: MFCD00000056


Purity: 99% CAS No.: 4165-57-5 Molecular Formula: C6D5Br Formula Weight: 162.06 Flash Point: 51°(124°F) Physical Form: Liquid Density: 1.55 Refractive Index: 1.557 Odor: Aromatic Boiling Point: 53°/23mm UN No.: 2514 MDL No.: MFCD00000056

Formula: C₆D₅Br
Boiling Pt: 53 °C (23 mmHg)
Melting Pt: –31 °C
Density: 1.55
Flash Pt: 51 °C (124 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000056
CAS Number: 4165-57-5
EINECS: 224-013-8
UN: 2514
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR