Acetonitrile-[D₃] (100% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

42266-KV 42266-SA
CAAA42266-KVEA 491.38 CAD
CAAA42266-KV CAAA42266-SA
Acetonitrile-[D₃] (100% D) (Isotopic) for NMR spectroscopy
Acetonitrile-[D3]

MDL: MFCD00001881 Notes: Minimum isotopic purity, 99.96%


Purity: 100% CAS No.: 2206-26-0 Molecular Formula: CD3CN Formula Weight: 44.08 Flash Point: 5°(42°F) Physical Form: Liquid in prescored ampoules, 0.75ml/ampoule Density: 0.844 Sensitivity: Hygroscopic Appearance: Colorless Odor: Pungent Boiling Point: 81° Merck Reference: 14,70 UN No.: 1648 MDL No.: MFCD00001881

Formula: D₃CCN
Boiling Pt: 81 °C
Melting Pt: –42 °C
Density: 0.844
Flash Pt: 5 °C (42 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II
Merck Index: 14,70

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR