Acetonitrile-[D₃] (99.8% D) (Isotopic) for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cyanomethane-D3, Acetonitrile-d3, Ethannitrile-D3, Trideuteroacetonitrile, Methyl-d3 cyanide, Methyl cyanide-D3, Acetonitrile-[²H₃]
42264-09
42264-18
CAAA42264-09EA
208.56
CAD
CAAA42264-09
CAAA42264-18
Acetonitrile-[D₃] (99.8% D) (Isotopic) for NMR spectroscopy
Acetonitrile-[D3]
MDL: MFCD00001881
Purity: 99.8%
CAS No.: 2206-26-0
Molecular Formula: CD3CN
Formula Weight: 44.08
Flash Point: 5°(42°F)
Physical Form: Liquid
Density: 0.844
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Pungent
Boiling Point: 81°
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001881
Formula:
D₃CCN MW: 44.03 g/mol Boiling Pt: 81 °C Melting Pt: –42 °C Density: 0.844 Flash Pt: 5 °C (42 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001881 CAS Number: 2206-26-0 EINECS: 218-616-5 UN: 1648 ADR: 3,II Merck Index: 14,70 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...