Aluminium ≥99.999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42259-09 42259-18 42259-30
CAAA42259-09EA 83.27 CAD
CAAA42259-09 CAAA42259-18 CAAA42259-30
Aluminium ≥99.999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.14g/slug Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR