Aluminium ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Aluminum
42258-18
42258-30
CAAA42258-09EA
96.44
CAD
CAAA42258-09
CAAA42258-18
CAAA42258-30
Aluminium ≥99.99% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Aluminium
MDL: MFCD00134029
Purity: 99.99%
CAS No.: 7429-90-5
Physical Form: ≈0.14g/slug
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2327 Melting Pt: 660 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...