Manganese(IV) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid
42250-30
42250-A1
CAAA42250-30EA
153.58
CAD
CAAA42250-30
CAAA42250-A1
Manganese(IV) oxide ≥99.9% (metals basis)
Manganese (IV) oxide
MDL: MFCD00003463
Purity: 99.9%
CAS No.: 1313-13-9
Molecular Formula: MnO2
Formula Weight: 86.94
Physical Form: -325 Mesh Powder
Density: 5.026
Appearance: Black
Melting Point: 535° dec.
Merck Reference: 14,5730
MDL No.: MFCD00003463
Formula:
MnO₂ MW: 86.94 g/mol Melting Pt: 535 (dec.) Density: 5.026 Storage Temperature: Ambient |
MDL Number:
MFCD00003463 CAS Number: 1313-13-9 EINECS: 215-202-6 Merck Index: 14,05730 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...