Silver perchlorate, anhydrous
Supplier: Thermo Scientific Chemicals
|
Danger
|
42209-06
42209-14
42209-22
CAAA42209-06EA
100.49
CAD
CAAA42209-06
CAAA42209-14
CAAA42209-22
Silver perchlorate, anhydrous
Silver perchlorate
MDL: MFCD00003400
Soluble in water and many organic solvents (benzene, toluene, pyridine)
CAS No.: 7783-93-9
Molecular Formula: AgClO4
Formula Weight: 207.32
Physical Form: Crystalline
Density: 2.806
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 485° dec.
Solubility: Soluble in water and many organic solvents (benzene, toluene, pyridine)
Merck Reference: 14,8523
UN No.: 1481
MDL No.: MFCD00003400
Formula:
AgClO₄ Melting Pt: 485 °C (dec.) Density: 2.806 Storage Temperature: Ambient |
MDL Number:
MFCD00003400 CAS Number: 7783-93-9 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II Merck Index: 14,8523 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...