Nickel chromium alloy (80:20 w%), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42195-G9
42195-H4
CAAA42195-G9EA
63.99
CAD
CAAA42195-G9
CAAA42195-H4
Nickel chromium alloy (80:20 w%), wire, Ø 0.25 mm (0.01 in)
Nickel Chromium alloy (80:20 w%)
MDL: MFCD00673110
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈0.43g/m
Odor: Odorless
MDL No.: MFCD00673110
Storage Temperature:
Ambient |
MDL Number:
MFCD00673110 CAS Number: 11106-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...