Bis(2-butoxyethyl)ether 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether
42150-AE
42150-AP
CAAA42150-ACEA
34.08
CAD
CAAA42150-AC
CAAA42150-AE
CAAA42150-AP
Bis(2-butoxyethyl)ether 99+%
Bis(2-butoxyethyl) ether
Extraction of precious metals
Purity: 99+%
CAS No.: 112-73-2
Molecular Formula: [CH3(CH2)3OCH2CH2]2O
Formula Weight: 218.74
Flash Point: 117°(243°F)
Physical Form: Liquid
Density: 0.885
Refractive Index: 1.4235
Appearance: Colorless
Melting Point: -60°
Boiling Point: 256°
MDL No.: MFCD00009459
Extraction of precious metals
Formula:
C₁₂H₂₆O₃ MW: 218.34 g/mol Boiling Pt: 256 Melting Pt: -60 Density: 0.885 Flash Pt: 117 °C (243 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009459 CAS Number: 112-73-2 EINECS: 204-001-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...