Tantalum ≥99.95% (metals basis), foil, Thickness: 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

42115-FF 42115-FI 42115-GH
CAAA42115-FFEA 237 CAD
CAAA42115-FF CAAA42115-FI CAAA42115-GH
Tantalum ≥99.95% (metals basis), foil, Thickness: 1.5 mm (0.06 in)
Tantalum

MDL: MFCD00011252


Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈15.6g/25x25mm Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR