Chromium(III) chloride hexahydrate ≥99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chromium trichloride hexahydrate, Chromic chloride hexahydrate

42114-22 42114-36
CAAA42114-22EA 122.61 CAD
CAAA42114-22 CAAA42114-36
Chromium(III) chloride hexahydrate ≥99.5%
Chromium (III) chloride hexahydrate

MDL: MFCD00149660


Purity: 99.5% min CAS No.: 10060-12-5 Molecular Formula: CrCl3.6H2O Formula Weight: 266.45 (158.35anhy) Physical Form: Crystalline Density: 1.76 Appearance: Dark green to black Odor: Odorless Melting Point: 83° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00149660

Formula: CrCl₃·6H₂O
MW: 266.45 g/mol
Melting Pt: 83 °C
Density: 1.76
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 14,2222

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR