Chromium(III) chloride hexahydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chromium trichloride hexahydrate, Chromic chloride hexahydrate
MDL: MFCD00149660
Slightly soluble in acetone. Soluble in water, alcohol. Insoluble in ether
Purity: 98%
CAS No.: 10060-12-5
Molecular Formula: CrCl3.6H2O
Formula Weight: 266.45 (158.35anhy)
Physical Form: Crystalline
Density: 1.835
Appearance: Dark Green
Odor: Odorless
Melting Point: 83°
Solubility: Slightly soluble in acetone. Soluble in water, alcohol. Insoluble in ether
Merck Reference: 14,2222
UN No.: 3260
MDL No.: MFCD00149660
Formula:
CrCl₃·6H₂O MW: 266.45 g/mol Melting Pt: 85 Density: 1.835 Storage Temperature: Ambient |
MDL Number:
MFCD00149660 CAS Number: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8 III Merck Index: 14,2222 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...