Lead(II) carbonate basic ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42089-36
42089-A3
CAAA42089-36EA
248.06
CAD
CAAA42089-36
CAAA42089-A3
Lead(II) carbonate basic ≥99% (metals basis)
Lead (II) carbonate basic
MDL: MFCD00078155
Soluble in acids, insoluble in water or alcohol
Purity: 99%
CAS No.: 1319-46-6
Molecular Formula: C2H2O8Pb3
Formula Weight: 775.63
Physical Form: -100 Mesh Powder
Density: 6.14
Melting Point: 400° dec.
Solubility: Soluble in acids, insoluble in water or alcohol
Merck Reference: 14,1005
UN No.: 3077
MDL No.: MFCD00078155
Formula:
(PbCO₃)₂·Pb(OH)₂ Melting Pt: 400 °C (dec.) Density: 6.14 Storage Temperature: Ambient |
MDL Number:
MFCD00078155 CAS Number: 1319-46-6 EINECS: 215-290-6 UN: 3077 ADR: 9,III Merck Index: 14,1005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...