Gallium(III) acetylacetonate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
42086-06
42086-14
CAAA42086-06EA
170.64
CAD
CAAA42086-06
CAAA42086-14
Gallium(III) acetylacetonate ≥99.99% (metals basis)
Gallium (III) acetylacetonate
MDL: MFCD00013492
Purity: 99.99%
CAS No.: 14405-43-7
Molecular Formula: C15H21O6Ga
Formula Weight: 367.05
Physical Form: Powder
Density: 1.42
Melting Point: 196-198° dec.
MDL No.: MFCD00013492
Formula:
C₁₅H₂₁GaO₆ Melting Pt: 196…198 °C (dec.) Density: 1.42 Storage Temperature: Ambient |
MDL Number:
MFCD00013492 CAS Number: 14405-43-7 EINECS: 238-377-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...