Petroleum spirit, 35…60 °C ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

42085-K2 42085-K7 42085-M1 42085-M4 42085-M6
CAAA42085-K2EA 94.64 CAD
CAAA42085-K2 CAAA42085-K7 CAAA42085-M1 CAAA42085-M4 CAAA42085-M6
Petroleum spirit, 35…60 °C ACS
Petroleum spirit

MDL: MFCD00081849


CAS No.: 8032-32-4 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.640 Refractive Index: 1.3630 Appearance: Colorless Odor: Petroleum-like Boiling Point: 35-60° UN No.: 1268 MDL No.: MFCD00081849

Boiling Pt: 35…60 °C
Density: 0.640
Flash Pt: –49 °C (–57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,I

Order Now

Specification Test Results

Color (APHA) 10
Boiling range 35-60°
Evaporation residue 0.001% max.
Acidity Pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR