HMDS (1,1,1,3,3,3-hexamethyldisilazane) 99+%, Electronic Grade for the electronics industry
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hexamethyldisilazane
MDL: MFCD00008259
Beilstein Registry No.: 635752
Notes: Low hydrolyzable chloride content
Fieser: 1,427 9,234 11,38 12,239 13,141 18,177 19,163 20,184 21,214
Purity: 99+%
CAS No.: 999-97-3
Molecular Formula: C6H19NSi2
Formula Weight: 161.40
Flash Point: 8°(46°F)
Physical Form: Liquid
Density: 0.774
Refractive Index: 1.4080
Sensitivity: Moisture Sensitive
Appearance: Colorless
Odor: Odorless
Melting Point: -78°
Boiling Point: 126°
Beilstein Reference: 635752
Merck Reference: 14,4689
UN No.: 3286
MDL No.: MFCD00008259
Formula:
(CH₃)₃SiNHSi(CH₃)₃ MW: 161.39 g/mol Boiling Pt: 126 Melting Pt: -82 Density: 0.774 Flash Pt: 8 °C (46 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008259 CAS Number: 999-97-3 EINECS: 213-668-5 UN: 3286 ADR: 3 II REACH: 01-2119438176-38 Merck Index: 14,4689 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...