Aluminium 99 Metals basis, spheres, Ø 2.38 mm (0.094 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

42029-KM 42029-KN
CAAA42029-KMEA 77.26 CAD
CAAA42029-KM CAAA42029-KN
Aluminium 99 Metals basis, spheres, Ø 2.38 mm (0.094 in)
Aluminium

MDL: MFCD00213972


Purity: 99% CAS No.: 7429-90-5 Molecular Formula: Alloy 1100, Al:Cu; 99+:0.12 wt% MDL No.: MFCD00213972

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR