Butyl laurate 99+%

Supplier: Thermo Scientific Chemicals

42024-AC 42024-AE 42024-AP
CAAA42024-ACEA 45.66 CAD
CAAA42024-AC CAAA42024-AE CAAA42024-AP
Butyl laurate 99+%
Butyl laurate

MDL: MFCD00042870 Beilstein Registry No.: 1776360


Purity: 99+% CAS No.: 106-18-3 Molecular Formula: CH3(CH2)10COO(CH2)3CH3 Formula Weight: 256.4 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.86 Refractive Index: 1.435 Melting Point: -10° Boiling Point: 166° Beilstein Reference: 1776360 MDL No.: MFCD00042870

Formula: C₁₆H₃₂O₂
Boiling Pt: 166 °C
Melting Pt: –10 °C
Density: 0,86
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042870
CAS Number: 106-18-3
EINECS: 203-370-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR