Antimony(V) chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

41978-18 41978-30
CAAA41978-18EA 113.76 CAD
CAAA41978-18 CAAA41978-30 CAAA41978-A1
Antimony(V) chloride 99%
Antimony (V) chloride

MDL: MFCD00011213 Notes: Aqueous solution hydrolyzes to Sb2O5 Soluble in HCl2 chloroform, carbon tetrachloride Fieser: 1,42 4,21 5,19 6,22 7,12 15,14 17,17 20,16


Purity: 99% CAS No.: 7647-18-9 Molecular Formula: SbCl5 Formula Weight: 299.02 Physical Form: Liquid Density: 2.336 Refractive Index: 1.601 Sensitivity: Moisture Sensitive Odor: Pungent Melting Point: 2.8° Boiling Point: 79°/22mm Solubility: Soluble in HCl, chloroform, carbon tetrachloride Merck Reference: 14,695 UN No.: 1730 MDL No.: MFCD00011213

Formula: Cl₅Sb
Boiling Pt: 79 °C (22 mmHg)
Melting Pt: 2.8 °C
Density: 2.336
Storage Temperature: Ambient
MDL Number: MFCD00011213
CAS Number: 7647-18-9
EINECS: 231-601-8
UN: 1730
ADR: 8,II
Merck Index: 14,695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR