Indium(III) chloride, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Indium Trichloride Chloride, Indium trichloride

41977-09 41977-18 41977-30
CAAA41977-09EA 155.63 CAD
CAAA41977-09 CAAA41977-18 CAAA41977-30
Indium(III) chloride, anhydrous ≥99.99% (metals basis)
Indium (III) chloride

MDL: MFCD00011058 Soluble in water Fieser: 16,181 19,173 20,197 21,228


Purity: 99.99% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Physical Form: Crystalline Density: 3.460 Appearance: White Odor: Odorless Melting Point: 586° Boiling Point: 800° Solubility: Soluble in water Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formula: Cl₃In
MW: 221.18 g/mol
Boiling Pt: 800 °C
Melting Pt: 586 °C
Density: 3.460
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8 II
Merck Index: 14,4958

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR