Potassium tellurite hydrate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41959-14
41959-22
CAAA41959-14EA
208.56
CAD
CAAA41959-14
CAAA41959-22
Potassium tellurite hydrate 97%
di-Potassium tellurium trioxide hydrate
Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.
Purity: 97%
CAS No.: 123333-66-4
Molecular Formula: K2O3Te.H2O
Formula Weight: 276.81 (253.80anhy)
Physical Form: Crystalline
Appearance: White
Melting Point: 465° dec.
UN No.: 3284
MDL No.: MFCD00149923
Formula:
K₂O₃Te·H₂O Melting Pt: 465 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149923 CAS Number: 123333-66-4 EINECS: 232-213-1 UN: 3284 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...