Zinc selenide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

41958-14 41958-22
CAAA41958-14EA 248.06 CAD
CAAA41958-14 CAAA41958-22
Zinc selenide ≥99.99% (metals basis)
Zinc selenide

MDL: MFCD00011296 Insoluble in water. Decomposes in dilute HNO3


Purity: 99.99% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: -325 Mesh Powder Density: 5.42 Melting Point: 1100° Solubility: Insoluble in water. Decomposes in dilute HNO3 Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formula: SeZn
Melting Pt: >1100 °C
Density: 5.42
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 14,10156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR