Molybdenum ≥99.5% (metals basis), spherical powder -170 mesh

Supplier: Thermo Scientific Chemicals
Warning

41919-18 41919-30 41919-A1
CAAA41919-18EA 54.51 CAD
CAAA41919-18 CAAA41919-30 CAAA41919-A1
Molybdenum ≥99.5% (metals basis), spherical powder -170 mesh
Molybdenum

MDL: MFCD00003465


Purity: 99.5% CAS No.: 7439-98-7 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1 III
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR