2,2,4-Trimethylpentane ≥99.5%, Environmental Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
Suitable for pesticide residue analysis
Purity: 99.5+%
CAS No.: 540-84-1
Molecular Formula: (CH3)2CHCH2C(CH3)3
Formula Weight: 114.23
Flash Point: -7°(19°F)
Physical Form: Liquid
Density: 0.692
Refractive Index: 1.3910
Odor: Nearly odorless
Melting Point: -107°
Boiling Point: 98-99°
Beilstein Reference: 1696876
Merck Reference: 14,5193
UN No.: 1262
MDL No.: MFCD00008943
Suitable for pesticide residue analysis
Formula:
(CH₃)₂CHCH₂C(CH₃)₃ Boiling Pt: 98…99 °C Melting Pt: –107 °C Density: 0.692 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008943 CAS Number: 540-84-1 EINECS: 208-759-1 UN: 1262 ADR: 3,II Merck Index: 14,5193 |
|
Specification Test Results
Color (APHA) | 10 |
Halogenated residue | 5ppt (GC-ECD) |
Evaporation residue | 3ppm |
H2O | 0.02% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...