(±)-2-Butanol, anhydrous 99%, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
Packaged under argon in resealable ChemSeal™ bottles.
MDL: MFCD00004569
Beilstein Registry No.: 1718765
Notes: H2O less than 0.01%
Purity: 99%
CAS No.: 78-92-2
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 26°(79°F)
Physical Form: Liquid
Density: 0.808
Refractive Index: 1.3970
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Sweet
Melting Point: -115°
Boiling Point: 98-101°
Beilstein Reference: 1718765
Merck Reference: 14,1541
UN No.: 1120
MDL No.: MFCD00004569
Formula:
H₃CCH₂CH(OH)CH₃ MW: 74.12 g/mol Boiling Pt: 98…101 °C Melting Pt: –115 °C Density: 0.808 Flash Pt: 26 °C (79 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004569 CAS Number: 78-92-2 EINECS: 201-158-5 UN: 1120 ADR: 3,II Merck Index: 14,1541 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...