1-Butanol, anhydrous ≥99.9%, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
Packaged under argon in resealable ChemSeal™ bottles.
MDL: MFCD00002964
Beilstein Registry No.: 969148
Notes: Water less than 0.01%
Purity: 99.9%
CAS No.: 71-36-3
Molecular Formula: CH3(CH2)3OH
Formula Weight: 74.12
Flash Point: 35°(95°F)
Physical Form: Liquid
Density: 0.810
Refractive Index: 1.3990
Sensitivity: Hygroscopic
Odor: Alcohol-like
pH: 7
Melting Point: -90 to -89°
Boiling Point: 117-118°
Beilstein Reference: 969148
Merck Reference: 14,1540
UN No.: 1120
MDL No.: MFCD00002964
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117…118 °C Melting Pt: –90…–89 °C Density: 0.810 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 14,1540 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...