Cetrimonium chloride ≥96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC
MDL: MFCD00011773
Beilstein Registry No.: 3657974
Purity: 96%
CAS No.: 112-02-7
Molecular Formula: C19H42ClN
Formula Weight: 320.00
Physical Form: Powder
Density: 0.968
Appearance: Off-white
Odor: Odorless
Melting Point: 234°
Beilstein Reference: 3657974
UN No.: 1759
MDL No.: MFCD00011773
Formula:
C₁₉H₄₂ClN MW: 320 g/mol Melting Pt: 234 °C Density: 0.968 Storage Temperature: Ambient |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 EINECS: 203-928-6 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...