Dicerium trisulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
41856-06
41856-14
CAAA41856-06EA
70.94
CAD
CAAA41856-06
CAAA41856-14
Dicerium trisulfide ≥99.9% (metals basis)
Dicerium trisulphide
MDL: MFCD00016007
Purity: 99.9%
CAS No.: 12014-93-6
Molecular Formula: Ce2S3
Formula Weight: 376.43
Physical Form: -325 Mesh Powder
Density: 5.02
Melting Point: 2100°
UN No.: 3134
MDL No.: MFCD00016007
Formula:
Ce₂S₃ Melting Pt: 2100 °C Density: 5,02 Storage Temperature: Ambient |
MDL Number:
MFCD00016007 CAS Number: 12014-93-6 EINECS: 234-603-7 UN: 3134 ADR: 4.3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...