1-Propanol, anhydrous ≥99.9%, chemSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane

Packaged under argon in resealable ChemSeal™ bottles.

41842-AK 41842-K2 41842-M1
CAAA41842-AKEA 52.3 CAD
CAAA41842-AK CAAA41842-K2 CAAA41842-M1
1-Propanol, anhydrous ≥99.9%, chemSeal™
1-Propanol

MDL: MFCD00002941 Beilstein Registry No.: 1098242 Notes: Water less than 100ppm


Purity: 99.9% CAS No.: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.10 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.804 Refractive Index: 1.3850 Odor: Alcohol-like Melting Point: -127° Boiling Point: 97° Beilstein Reference: 1098242 Merck Reference: 14,7842 UN No.: 1274 MDL No.: MFCD00002941

Formula: H₃CCH₂CH₂OH
MW: 60.1 g/mol
Boiling Pt: 97
Melting Pt: -127
Density: 0.804
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 14,7842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR