Copper(I) chloride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cuprous chloride, Copper chloride
41837-22
41837-36
CAAA41837-22EA
216.46
CAD
CAAA41837-22
CAAA41837-36
Copper(I) chloride ≥99% (metals basis)
Copper (I) chloride
MDL: MFCD00010971
Purity: 99%
CAS No.: 7758-89-6
Molecular Formula: CuCl
Formula Weight: 98.99
Physical Form: -80 Mesh Powder
Density: 4.14
Refractive Index: 1.93
Sensitivity: Air and Moisture Sensitive
Appearance: White
Melting Point: 430°
Boiling Point: 1490°
Merck Reference: 14,2660
UN No.: 2802
MDL No.: MFCD00010971
Formula:
ClCu MW: 99 g/mol Boiling Pt: 1367 Melting Pt: 430 Density: 4.14 Storage Temperature: Ambient |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8 III Merck Index: 14,2660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...