Lithium tantalate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium tantalum trioxide, Lithium tantalum oxide

41834-09 41834-18
CAAA41834-09EA 202.24 CAD
CAAA41834-09 CAAA41834-18
Lithium tantalate ≥99.9% (metals basis)
Lithium tantalate

MDL: MFCD00016174


Purity: 99.9% CAS No.: 12031-66-2 Molecular Formula: LiTaO3 Formula Weight: 235.89 Physical Form: -200 Mesh Powder Odor: Odorless MDL No.: MFCD00016174

Formula: LiO₃Ta
MW: 235.89
Storage Temperature: Ambient
MDL Number: MFCD00016174
CAS Number: 12031-66-2
EINECS: 234-757-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR