Dilithium oxide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

41832-09 41832-18 41832-30
CAAA41832-09EA 123.4 CAD
CAAA41832-09 CAAA41832-18 CAAA41832-30
Dilithium oxide ≥99.5% (metals basis)
di-Lithium oxide

Ceramics and special glass formulations, carbon dioxide absorbent


Purity: 99.5% CAS No.: 12057-24-8 Molecular Formula: Li2O Formula Weight: 29.88 Physical Form: -100 Mesh Powder Density: 2.013 Melting Point: 1427° Merck Reference: 14,5538 UN No.: 3262 MDL No.: MFCD00016183


Ceramics and special glass formulations, carbon dioxide absorbent

Formula: Li₂O
Melting Pt: 1427 °C
Density: 2.013
Storage Temperature: Ambient
MDL Number: MFCD00016183
CAS Number: 12057-24-8
EINECS: 235-019-5
UN: 3262
ADR: 8,II
Merck Index: 14,5538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR