Molybdenum(IV) sulfide ≥98% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Molybdenum disulfide, Molybdenum disulphide

41827-14 41827-22 41827-36
CAAA41827-14EA 44.24 CAD
CAAA41827-14 CAAA41827-22 CAAA41827-36
Molybdenum(IV) sulfide ≥98% (metals basis)
Molybdenum (IV) sulphide

Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.


  • -325 Mesh Powder


Purity: 99% CAS No.: 1317-33-5 Molecular Formula: MoS2 Formula Weight: 160.08 Physical Form: -325 Mesh Powder Appearance: Black Odor: Odorless Melting Point: 1185° Merck Reference: 14,6236 MDL No.: MFCD00003470

Formula: MoS₂
MW: 160.07 g/mol
Melting Pt: 1185 °C
Density: 4.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 14,6236

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR